3-Fluoro-4-[3-(trifluoromethyl)-1H-pyrazol-1-yl]aniline

Lieferant: Apollo Scientific

PC410472-1G
APOSPC410472-1GEA 647 EUR
APOSPC410472-1G
3-Fluoro-4-[3-(trifluoromethyl)-1H-pyrazol-1-yl]aniline
3-Fluoro-4-[3-(trifluoromethyl)-1H-pyrazol-1-yl]aniline
Formel: C₁₀H₇F₄N₃
Molecular Weight: 245,176 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08699601
CAS-Nummer: 1006468-65-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR