3-[4-Chloro-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
Lieferant: Apollo Scientific
PC410473-1G
APOSPC410473-1GEA
0
EUR
APOSPC410473-1G
3-[4-Chloro-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
3-[4-Chloro-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
Formel:
C₇H₆ClF₃N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD04969243 CAS-Nummer: 1006470-23-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...