2-[4-Chloro-5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid

Lieferant: Apollo Scientific

PC410474-1G
APOSPC410474-1GEA 648 EUR
APOSPC410474-1G
2-[4-Chloro-5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
2-[4-Chloro-5-cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanoic acid
Formel: C₁₀H₁₀ClF₃N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03419746
CAS-Nummer: 1006470-39-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR