4-Bromo-1-(2,2,2-trifluoroethyl)-1H-pyrazol-3-amine

Lieferant: Apollo Scientific
Achtung

PC410475-1G
APOSPC410475-1GEA 980 EUR
APOSPC410475-1G
4-Bromo-1-(2,2,2-trifluoroethyl)-1H-pyrazol-3-amine
4-Bromo-1-(2,2,2-trifluoroethyl)-1H-pyrazol-3-amine
Formel: C₅H₅BrF₃N₃
Molecular Weight: 244,013 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08696329
CAS-Nummer: 1006471-06-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR