[4-Chloro-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetonitrile

Lieferant: Apollo Scientific

PC410476-1G
APOSPC410476-1GEA 516 EUR
APOSPC410476-1G
[4-Chloro-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetonitrile
[4-Chloro-3-(trifluoromethyl)-1H-pyrazol-1-yl]acetonitrile
Formel: C₆H₃ClF₃N₃
Molecular Weight: 209,556 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04969710
CAS-Nummer: 1006471-49-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR