1-(2,3-Difluorophenyl)-N-[(1-methyl-1H-pyrazol-4-yl)methyl]methanamine

Lieferant: Apollo Scientific

PC410477-1G
APOSPC410477-1GEA 828 EUR
APOSPC410477-1G
1-(2,3-Difluorophenyl)-N-[(1-methyl-1H-pyrazol-4-yl)methyl]methanamine
1-(2,3-Difluorophenyl)-N-[(1-methyl-1H-pyrazol-4-yl)methyl]methanamine
Formel: C₁₂H₁₃F₂N₃
Lagertemperatur: Raumtemperatur
MDL: MFCD08696782
CAS-Nummer: 1006472-90-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR