3,5-Dimethyl-1-[(2,2,2-trifluoroethoxy)methyl]-1H-pyrazol-4-amine

Lieferant: Apollo Scientific

PC410478-1G
APOSPC410478-1GEA 647 EUR
APOSPC410478-1G
3,5-Dimethyl-1-[(2,2,2-trifluoroethoxy)methyl]-1H-pyrazol-4-amine
3,5-Dimethyl-1-[(2,2,2-trifluoroethoxy)methyl]-1H-pyrazol-4-amine
Formel: C₈H₁₂F₃N₃O
Molecular Weight: 223,196 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04970047
CAS-Nummer: 1006473-39-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR