2-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid

Lieferant: Apollo Scientific

PC410479-1G
APOSPC410479-1GEA 647 EUR
APOSPC410479-1G
2-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid
2-[3-(Trifluoromethyl)-1H-pyrazol-1-yl]butanoic acid
Formel: C₈H₉F₃N₂O₂
Molecular Weight: 222,164 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04970063
CAS-Nummer: 1006473-54-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR