1-(3,5-Difluorophenyl)-N-[(1,5-dimethyl-1H-pyrazol-4-yl)methyl]methanamine

Lieferant: Apollo Scientific

PC410480-1G
APOSPC410480-1GEA 828 EUR
APOSPC410480-1G
1-(3,5-Difluorophenyl)-N-[(1,5-dimethyl-1H-pyrazol-4-yl)methyl]methanamine
1-(3,5-Difluorophenyl)-N-[(1,5-dimethyl-1H-pyrazol-4-yl)methyl]methanamine
Formel: C₁₃H₁₅F₂N₃
Molecular Weight: 251,275 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08696831
CAS-Nummer: 1006473-57-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR