2-[4-Chloro-3-(trifluoromethyl)-1H-pyrazol-1-yl]ethanamine
Lieferant: Apollo Scientific
PC410482-1G
APOSPC410482-1GEA
828
EUR
APOSPC410482-1G
2-[4-Chloro-3-(trifluoromethyl)-1H-pyrazol-1-yl]ethanamine
2-[4-Chloro-3-(trifluoromethyl)-1H-pyrazol-1-yl]ethanamine
Formel:
C₆H₇ClF₃N₃ Molecular Weight: 213,588 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04970068 CAS-Nummer: 1006473-61-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...