[6-(1,3-Dimethyl-1H-pyrazol-4-yl)-3-methyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid

Lieferant: Apollo Scientific

PC410483-1G
APOSPC410483-1GEA 828 EUR
APOSPC410483-1G
[6-(1,3-Dimethyl-1H-pyrazol-4-yl)-3-methyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
[6-(1,3-Dimethyl-1H-pyrazol-4-yl)-3-methyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
Formel: C₁₅H₁₄F₃N₅O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08697653
CAS-Nummer: 1006477-40-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR