[6-(1,5-Dimethyl-1H-pyrazol-4-yl)-3-methyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid

Lieferant: Apollo Scientific

PC410484-1G
APOSPC410484-1GEA 980 EUR
APOSPC410484-1G
[6-(1,5-Dimethyl-1H-pyrazol-4-yl)-3-methyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
[6-(1,5-Dimethyl-1H-pyrazol-4-yl)-3-methyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
Formel: C₁₅H₁₄F₃N₅O₂
Molecular Weight: 353,299 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08697655
CAS-Nummer: 1006477-44-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR