[3-Cyclopropyl-6-(1-ethyl-1H-pyrazol-4-yl)-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid

Lieferant: Apollo Scientific

PC410485-1G
APOSPC410485-1GEA 828 EUR
APOSPC410485-1G
[3-Cyclopropyl-6-(1-ethyl-1H-pyrazol-4-yl)-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
Formel: C₁₇H₁₆F₃N₅O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08697672
CAS-Nummer: 1006477-52-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR