[4-(Difluoromethyl)-6-(1,5-dimethyl-1H-pyrazol-4-yl)-3-methyl-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid

Lieferant: Apollo Scientific

PC410486-1G
APOSPC410486-1GEA 980 EUR
APOSPC410486-1G
[4-(Difluoromethyl)-6-(1,5-dimethyl-1H-pyrazol-4-yl)-3-methyl-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
[4-(Difluoromethyl)-6-(1,5-dimethyl-1H-pyrazol-4-yl)-3-methyl-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
Formel: C₁₅H₁₅F₂N₅O₂
Molecular Weight: 335,309 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08697730
CAS-Nummer: 1006477-98-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR