[3-Cyclopropyl-4-(difluoromethyl)-6-(1,5-dimethyl-1H-pyrazol-4-yl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid

Lieferant: Apollo Scientific

PC410487-1G
APOSPC410487-1GEA 828 EUR
APOSPC410487-1G
[3-Cyclopropyl-4-(difluoromethyl)-6-(1,5-dimethyl-1H-pyrazol-4-yl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
[3-Cyclopropyl-4-(difluoromethyl)-6-(1,5-dimethyl-1H-pyrazol-4-yl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
Formel: C₁₇H₁₇F₂N₅O₂
Molecular Weight: 361,346 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08697750
CAS-Nummer: 1006478-10-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR