[3-Cyclopropyl-4-(difluoromethyl)-6-(1-ethyl-5-methyl-1H-pyrazol-4-yl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid

Lieferant: Apollo Scientific

PC410488-1G
APOSPC410488-1GEA 828 EUR
APOSPC410488-1G
[3-Cyclopropyl-4-(difluoromethyl)-6-(1-ethyl-5-methyl-1H-pyrazol-4-yl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
[3-Cyclopropyl-4-(difluoromethyl)-6-(1-ethyl-5-methyl-1H-pyrazol-4-yl)-1H-pyrazolo[3,4-b]pyridin-1-yl]acetic acid
Formel: C₁₈H₁₉F₂N₅O₂
Molecular Weight: 375,373 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08697757
CAS-Nummer: 1006478-14-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR