2-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]ethanamine

Lieferant: Apollo Scientific

PC410489-1G
APOSPC410489-1GEA 980 EUR
APOSPC410489-1G
2-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]ethanamine
2-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]ethanamine
Formel: C₉H₁₂F₃N₃
Molecular Weight: 219,207 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06804826
CAS-Nummer: 1006479-50-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR