2-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]ethanol

Lieferant: Apollo Scientific

PC410490-1G
APOSPC410490-1GEA 647 EUR
APOSPC410490-1G
2-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]ethanol
2-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]ethanol
Formel: C₉H₁₁F₃N₂O
Molecular Weight: 220,192 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06804951
CAS-Nummer: 1006480-18-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR