3-[4-Chloro-3-(trifluoromethyl)-1H-pyrazol-1-yl]propan-1-amine
Lieferant: Apollo Scientific
PC410491-1G
APOSPC410491-1GEA
647
EUR
APOSPC410491-1G
3-[4-Chloro-3-(trifluoromethyl)-1H-pyrazol-1-yl]propan-1-amine
3-[4-Chloro-3-(trifluoromethyl)-1H-pyrazol-1-yl]propan-1-amine
Formel:
C₇H₉ClF₃N₃ Molecular Weight: 227,615 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04970070 CAS-Nummer: 1006482-40-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...