1-{[1-(Difluoromethyl)-5-methyl-1H-pyrazol-4-yl]sulphonyl}piperazine

Lieferant: Apollo Scientific

PC410493-1G
APOSPC410493-1GEA 828 EUR
APOSPC410493-1G
1-{[1-(Difluoromethyl)-5-methyl-1H-pyrazol-4-yl]sulphonyl}piperazine
1-{[1-(Difluoromethyl)-5-methyl-1H-pyrazol-4-yl]sulphonyl}piperazine
Formel: C₉H₁₄F₂N₄O₂S
Molecular Weight: 280.295 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04970132
CAS-Nummer: 1006482-97-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR