1-(2,2-Difluoroethyl)-1H-pyrazole-3-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Carboxy-1-(2,2-difluoroethyl)-1H-pyrazole
PC410494-1G
APOSPC410494-1GEA
828
EUR
APOSPC410494-1G
1-(2,2-Difluoroethyl)-1H-pyrazole-3-carboxylic acid
1-(2,2-Difluoroethyl)-1H-pyrazole-3-carboxylic acid
Formel:
C₆H₆F₂N₂O₂ Molecular Weight: 176,121 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08696509 CAS-Nummer: 1006486-39-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...