2-{1-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propan-2-yl}-4-methyl-1,3-thiazole-5-carboxylic acid

Lieferant: Apollo Scientific

PC410495-1G
APOSPC410495-1GEA 386 EUR
APOSPC410495-1G
2-{1-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propan-2-yl}-4-methyl-1,3-thiazole-5-carboxylic acid
2-{1-[5-Cyclopropyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propan-2-yl}-4-methyl-1,3-thiazole-5-carboxylic acid
Formel: C₁₅H₁₆F₃N₃O₂S
Molecular Weight: 359,367 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08558261
CAS-Nummer: 1006682-82-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR