3-Cyclopropyl-1-(2,2,2-trifluoroethyl)-1H-pyrazole-4-carboxylic acid

Lieferant: Apollo Scientific

PC410496-1G
APOSPC410496-1GEA 0 EUR
APOSPC410496-1G
3-Cyclopropyl-1-(2,2,2-trifluoroethyl)-1H-pyrazole-4-carboxylic acid
3-Cyclopropyl-1-(2,2,2-trifluoroethyl)-1H-pyrazole-4-carboxylic acid
Formel: C₉H₉F₃N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08696531
CAS-Nummer: 1006486-60-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR