5-Methyl-1-(2,2,2-trifluoroethyl)-1H-pyrazol-3-amine

Lieferant: Apollo Scientific

PC410497-1G
APOSPC410497-1GEA 516 EUR
APOSPC410497-1G
5-Methyl-1-(2,2,2-trifluoroethyl)-1H-pyrazol-3-amine
5-Methyl-1-(2,2,2-trifluoroethyl)-1H-pyrazol-3-amine
Formel: C₆H₈F₃N₃
Molecular Weight: 179,143 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08696547
CAS-Nummer: 1006486-85-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR