1-(2,2-Difluoroethyl)-5-methyl-1H-pyrazol-3-amine
Lieferant: Apollo Scientific
PC410498-1G
APOSPC410498-1GEA
0
EUR
APOSPC410498-1G
1-(2,2-Difluoroethyl)-5-methyl-1H-pyrazol-3-amine
1-(2,2-Difluoroethyl)-5-methyl-1H-pyrazol-3-amine
Formel:
C₆H₉F₂N₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08696549 CAS-Nummer: 1006486-89-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...