4-Chloro-1-[(3-fluorophenoxy)methyl]-1H-pyrazole-3-carboxylic acid

Lieferant: Apollo Scientific

PC410499-1G
APOSPC410499-1GEA 232 EUR
APOSPC410499-1G
4-Chloro-1-[(3-fluorophenoxy)methyl]-1H-pyrazole-3-carboxylic acid
4-Chloro-1-[(3-fluorophenoxy)methyl]-1H-pyrazole-3-carboxylic acid
Formel: C₁₁H₈ClFN₂O₃
Molecular Weight: 270,644 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04968048
CAS-Nummer: 1006489-14-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR