2-Bromo-1-(3-fluoro-3-methyl-azetidin-1-yl)ethanone

Lieferant: Apollo Scientific

PC411260-1G
APOSPC411260-1GEA 287 EUR
APOSPC411260-1G
2-Bromo-1-(3-fluoro-3-methyl-azetidin-1-yl)ethanone
2-Bromo-1-(3-fluoro-3-methyl-azetidin-1-yl)ethanone
Formel: C₆H₉BrFNO
Molecular Weight: 210,044 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31697597
CAS-Nummer: 1466514-89-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR