3-(Fluoromethyl)azetidine hydrochloride

Lieferant: Apollo Scientific

PC411261-1G PC411261-250MG
APOSPC411261-1GEA 0 EUR
APOSPC411261-1G APOSPC411261-250MG
3-(Fluoromethyl)azetidine hydrochloride
3-(Fluoromethyl)azetidine hydrochloride
Formel: C₄H₈FN·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD27991715
CAS-Nummer: 1642298-59-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR