2-(Fluoromethyl)azetidine trifluoroacetic acid

Lieferant: Apollo Scientific

PC411262-1G
APOSPC411262-1GEA 1610 EUR
APOSPC411262-1G
2-(Fluoromethyl)azetidine trifluoroacetic acid
2-(Fluoromethyl)azetidine trifluoroacetic acid
Formel: C₄H₈FN·CF₃CO₂H
Molecular Weight: 203,135 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31696003
CAS-Nummer: 2227206-76-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR