(2S)-2-(Fluoromethyl)azetidine trifluoroacetic acid
Lieferant: Apollo Scientific
PC411263-1G
APOSPC411263-1GEA
1780
EUR
APOSPC411263-1G
(2S)-2-(Fluoromethyl)azetidine trifluoroacetic acid
(2S)-2-(Fluoromethyl)azetidine trifluoroacetic acid
Formel:
C₄H₈FN·CF₃CO₂H Molecular Weight: 203,135 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD31620804 CAS-Nummer: 2173637-40-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...