3-(Difluoromethyl)-3-methylazetidine hydrochloride

Lieferant: Apollo Scientific

PC411265-1G
APOSPC411265-1GEA 2050 EUR
APOSPC411265-1G
3-(Difluoromethyl)-3-methylazetidine hydrochloride
3-(Difluoromethyl)-3-methylazetidine hydrochloride
Formel: C₅H₉F₂N·ClH
Molecular Weight: 157,589 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31696021
CAS-Nummer: 2227206-51-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR