3-(Trifluoromethoxy)azetidine hydrochloride

Lieferant: Apollo Scientific

PC411266-1G
APOSPC411266-1GEA 1830 EUR
APOSPC411266-1G
3-(Trifluoromethoxy)azetidine hydrochloride
3-(Trifluoromethoxy)azetidine hydrochloride
Formel: C₄H₆F₃NO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD26097230
CAS-Nummer: 1803590-72-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR