2-(3,3-Difluoroazetidin-1-yl)ethan-1-amine

Lieferant: Apollo Scientific

PC411267-1G
APOSPC411267-1GEA 1710 EUR
APOSPC411267-1G
2-(3,3-Difluoroazetidin-1-yl)ethan-1-amine
2-(3,3-Difluoroazetidin-1-yl)ethan-1-amine
Formel: C₅H₁₀F₂N₂
Molecular Weight: 136,143 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD29994892
CAS-Nummer: 2045189-46-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR