3-Fluorocyclobutan-1-one

Lieferant: Apollo Scientific

PC411268-1G
APOSPC411268-1GEA 3380 EUR
APOSPC411268-1G
3-Fluorocyclobutan-1-one
3-Fluorocyclobutan-1-one
Formel: C₄H₅FO
Lagertemperatur: Raumtemperatur
MDL: MFCD23106394
CAS-Nummer: 1415342-84-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR