(2,2-Difluorocyclobutyl)methanamine hydrochloride

Lieferant: Apollo Scientific

PC411269-1G
APOSPC411269-1GEA 2500 EUR
APOSPC411269-1G
(2,2-Difluorocyclobutyl)methanamine hydrochloride
(2,2-Difluorocyclobutyl)methanamine hydrochloride
Formel: C₅H₉F₂N·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD30530422
CAS-Nummer: 2055839-95-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR