3-(Trifluoromethyl)cyclobutan-1-amine hydrochloride

Lieferant: Apollo Scientific

PC411274-1G
APOSPC411274-1GEA 2890 EUR
APOSPC411274-1G
3-(Trifluoromethyl)cyclobutan-1-amine hydrochloride
3-(Trifluoromethyl)cyclobutan-1-amine hydrochloride
Formel: C₅H₈F₃N·ClH
Molecular Weight: 175.58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28714563
CAS-Nummer: 1803601-06-0

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR