cis-3-Fluoro-3-methylcyclobutane-1-carboxylic acid
Lieferant: Apollo Scientific
PC411276-1G
APOSPC411276-1GEA
2450
EUR
APOSPC411276-1G
cis-3-Fluoro-3-methylcyclobutane-1-carboxylic acid
cis-3-Fluoro-3-methylcyclobutane-1-carboxylic acid
Formel:
C₆H₉FO₂ Molecular Weight: 132,133 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD29477491 CAS-Nummer: 1455037-45-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...