trans-3-Fluorocyclobutanol

Lieferant: Apollo Scientific

PC411277-1G
APOSPC411277-1GEA 1520 EUR
APOSPC411277-1G
trans-3-Fluorocyclobutanol
trans-3-Fluorocyclobutanol
Formel: C₄H₇FO
Molecular Weight: 90.096 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20489133
CAS-Nummer: 1262278-60-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR