3-Fluoro-1-methylcyclobutan-1-amine hydrochloride

Lieferant: Apollo Scientific

PC411281-1G
APOSPC411281-1GEA 1490 EUR
APOSPC411281-1G
3-Fluoro-1-methylcyclobutan-1-amine hydrochloride
3-Fluoro-1-methylcyclobutan-1-amine hydrochloride
Formel: C₅H₁₀FN·ClH
Molecular Weight: 139,599 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31618008
CAS-Nummer: 1781122-54-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR