7-Fluoro-5-azaspiro[2.4]heptane hydrochloride

Lieferant: Apollo Scientific

PC411283-1G
APOSPC411283-1GEA 1610 EUR
APOSPC411283-1G
7-Fluoro-5-azaspiro[2.4]heptane hydrochloride
7-Fluoro-5-azaspiro[2.4]heptane hydrochloride
Formel: C₆H₁₀FN·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD31697808
CAS-Nummer: 2227205-26-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR