6-Fluoro-2-azaspiro[3.3]heptane trifluoroacetic acid

Lieferant: Apollo Scientific

Synonyme: 6-Fluoro-2-azaspiro[3.3]heptane 2,2,2-trifluoroacetate

PC411284-1G
APOSPC411284-1GEA 659 EUR
APOSPC411284-1G
6-Fluoro-2-azaspiro[3.3]heptane trifluoroacetic acid
6-Fluoro-2-azaspiro[3.3]heptane trifluoroacetic acid
Formel: C₆H₁₀FN·CF₃CO₂H
Molecular Weight: 229.172 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31695984
CAS-Nummer: 2227205-88-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR