1,1-Difluorospiro[2.5]octan-6-one

Lieferant: Apollo Scientific

PC411288-1G
APOSPC411288-1GEA 0 EUR
APOSPC411288-1G
1,1-Difluorospiro[2.5]octan-6-one
1,1-Difluorospiro[2.5]octan-6-one
Formel: C₈H₁₀F₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD28126838
CAS-Nummer: 1513853-81-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR