1-(Difluoromethyl)-4-iodo-3-methyl-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

PC411302-1G PC411302-5G
APOSPC411302-1GEA 33.5 EUR
APOSPC411302-1G APOSPC411302-5G
1-(Difluoromethyl)-4-iodo-3-methyl-1H-pyrazole
1-(Difluoromethyl)-4-iodo-3-methyl-1H-pyrazole
Formel: C₅H₅F₂IN₂
Lagertemperatur: Kühlschrank
MDL: MFCD15146436
CAS-Nummer: 1217862-89-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR