1-(2-Fluoroethyl)-4-iodo-3-methyl-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

PC411305-1G PC411305-5G
APOSPC411305-1GEA 30.6 EUR
APOSPC411305-1G APOSPC411305-5G
1-(2-Fluoroethyl)-4-iodo-3-methyl-1H-pyrazole
1-(2-Fluoroethyl)-4-iodo-3-methyl-1H-pyrazole
Formel: C₆H₈FIN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD25949444
CAS-Nummer: 1443278-85-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR