2-(Difluoromethoxy)-5-fluoroaniline hydrochloride

Lieferant: Apollo Scientific
Achtung

PC411308-1G PC411308-5G
APOSPC411308-1GEA 36.4 EUR
APOSPC411308-1G APOSPC411308-5G
2-(Difluoromethoxy)-5-fluoroaniline hydrochloride
2-(Difluoromethoxy)-5-fluoroaniline hydrochloride
Formel: C₇H₆F₃NO·ClH
Molecular Weight: 213.585 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD25368949
CAS-Nummer: 1431966-19-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR