5-Chloro-2-(difluoromethoxy)aniline hydrochloride

Lieferant: Apollo Scientific
Achtung

PC411311-1G PC411311-5G
APOSPC411311-1GEA 34.8 EUR
APOSPC411311-1G APOSPC411311-5G
5-Chloro-2-(difluoromethoxy)aniline hydrochloride
5-Chloro-2-(difluoromethoxy)aniline hydrochloride
Formel: C₇H₆ClF₂NO·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD25371274
CAS-Nummer: 1431963-04-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR