1-Difluoromethyl-1H-pyrazole-3-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
PC411317-1G
PC411317-25G
PC411317-5G
APOSPC411317-1GEA
0
EUR
APOSPC411317-1G
APOSPC411317-25G
APOSPC411317-5G
1-Difluoromethyl-1H-pyrazole-3-carboxylic acid
1-Difluoromethyl-1H-pyrazole-3-carboxylic acid
Formel:
C₅H₄F₂N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03420211 CAS-Nummer: 925179-02-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...