4-Bromo-1-(difluoromethyl)-3-methyl-1H-pyrazole

Lieferant: Apollo Scientific
Achtung

PC411325-1G PC411325-25G PC411325-5G
APOSPC411325-1GEA 62.5 EUR
APOSPC411325-1G APOSPC411325-25G APOSPC411325-5G
4-Bromo-1-(difluoromethyl)-3-methyl-1H-pyrazole
4-Bromo-1-(difluoromethyl)-3-methyl-1H-pyrazole
Formel: C₅H₅BrF₂N₂
Molecular Weight: 211.01 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14702889
CAS-Nummer: 1215295-92-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR