(2-Fluorphenyl)essigsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(2-Fluorophenyl)acetic acid, 2-Fluorophenylacetic acid

PC4120-100G
APOSPC4120-100GEA 31.9 EUR
APOSPC4120-100G
(2-Fluorphenyl)essigsäure 98%
(2-Fluorphenyl)essigsäure
Formel: (C₆H₄F)CH₂COOH
Molecular Weight: 154,14 g/mol
Schmelzpunkt: 61…65 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004315
CAS-Nummer: 451-82-1
EINECS: 207-196-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR