2-Chlor-6-fluor-5-methylphenylboronsäure 97%

Lieferant: Apollo Scientific

PC412003-5G
APOSPC412003-5GEA 65.4 EUR
APOSPC412003-5G
2-Chlor-6-fluor-5-methylphenylboronsäure 97%
2-Chlor-6-fluor-5-methylphenylboronsäure
Formel: C₇H₇BClFO₂
Molecular Weight: 188,3917 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03701535
CAS-Nummer: 352535-86-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR